Assay Details
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011942.1 | NP_036072.1 | 1 - 2 | 78 | 91 | |
| GenBank mRNA | AB009653.1 | - | 1 - 2 | 78 | 91 | |
| AK075590.1 | - | 1 - 2 | 70 | 91 | ||
| AK089112.1 | - | 1 - 2 | 70 | 91 | ||
| AK167478.1 | - | 1 - 2 | 65 | 91 | ||
| BC068120.1 | - | 1 - 2 | 104 | 5714650 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB009653.1,AK075590.1,AK089112.1,AK167478.1,BC068120.1,NM_011942.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
lipid metabolic process
fatty acid metabolic process
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331511 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331513 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186572 | AK167478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186572 | BC068120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186572 | AB009653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186572 | AK003689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186572 | AK075590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966709 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110805 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186572 | AK167478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186572 | BC068120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186572 | AB009653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186572 | AK003689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186572 | AK075590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00997269 | - | - |
| Whole Mouse Genome G4122A | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520084 | AB009653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520084 | AK075590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520084 | NM_011942.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106249 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489479 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489473 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03516971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03489489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331511 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03676417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03489489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03776466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517435 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417433_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517425 | - | - |
| Mouse Genome 430 2.0 Array | 1417433_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417433_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26394_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26394_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400031105.mm.1 | NM_011942.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031105.mm.1 | NM_011942.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031105.mm.1 | TC0400003775.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031102.mm.1 | NM_011942.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031102.mm.1 | NM_011942.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031102.mm.1 | TC0400003775.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417433_PM_at | - | - |
| Mouse Expression Set 430 | 1417433_at | - | - |
| Murine Genome U74v2 Set | 98429_at | - | - |
| Murine Genome U74v2 Set | 98429_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417433_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417433_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417433_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417433_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 932099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1500112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228132 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228132 | - | - |
| mouse-6 v1.1 (Illumina) | scl23666.9_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23666.9_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228132 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228132 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23666.9_53-S | - | - |

Lypla2
Genomic Map