Assay Details
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011942.1 | NP_036072.1 | 8 - 9 | 574 | 102 | |
| GenBank mRNA | AB009653.1 | - | 8 - 9 | 574 | 102 | |
| AK003689.1 | - | 8 - 9 | 541 | 102 | ||
| AK075590.1 | - | 8 - 9 | 566 | 102 | ||
| AK089112.1 | - | 8 - 9 | 566 | 102 | ||
| AK167478.1 | - | 8 - 9 | 561 | 102 | ||
| BC068120.1 | - | 8 - 9 | 600 | 5714650 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypla2
Genomic Map