Assay Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013536.2 | NP_038564.1 | 2 - 3 | 407 | 138 | |
| GenBank mRNA | AK081757.1 | - | 2 - 3 | 385 | 138 | |
| BC002004.1 | - | 2 - 3 | 327 | 3487480 | 138 | |
| CB948801.1 | - | 2 - 3 | 410 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emg1
Genomic Map