Assay Details
| Gene Symbol: Zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013705.1 | NP_038733.1 | 2 - 3 | 204 | 81 | |
| GenBank mRNA | AY605122.1 | - | 2 - 3 | 204 | 81 | |
| BC049120.1 | - | 2 - 3 | 205 | 6813126 | 81 | |
| BC051972.1 | - | 2 - 3 | 404 | 5707652 | 81 | |
| Z30174.1 | - | 2 - 3 | 204 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp30
Genomic Map