Assay Details
| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013711.3 | NP_038739.2 | 3 - 4 | 262 | 56 | |
| GenBank mRNA | AB027566.1 | - | 3 - 4 | 262 | 56 | |
| AF136399.1 | - | 3 - 4 | 265 | 56 | ||
| AF171053.1 | - | 3 - 4 | 228 | 56 | ||
| AF412308.1 | - | 3 - 4 | 264 | 56 | ||
| AK008004.1 | - | 3 - 4 | 181 | 56 | ||
| AK075828.1 | - | 3 - 4 | 240 | 56 | ||
| BC013688.1 | - | 3 - 4 | 242 | 3584154 | 56 | |
| BC029344.1 | - | 3 - 4 | 242 | 56 | ||
| BC052157.1 | - | 3 - 4 | 241 | 6506286 | 56 | |
| BC052758.1 | - | 3 - 4 | 241 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd2
Genomic Map