Assay Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF037043.1 | - | 2 - 3 | 199 | 84 | |
| AF046887.1 | - | 2 - 3 | 276 | 84 | ||
| BC021598.1 | - | 1 - 2 | 177 | 5098857 | 84 | |
| CF106558.1 | - | 2 - 3 | 212 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpmt
Genomic Map