Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 9 - 10 | 1490 | 76 | |
| NM_001301688.1 | NP_001288617.1 | 8 - 9 | 1330 | 76 | ||
| NM_018745.5 | NP_061215.1 | 8 - 9 | 1330 | 76 | ||
| NR_125913.1 | 8 - 9 | 1482 | 76 | |||
| GenBank mRNA | AF032128.1 | - | 6 - 7 | 756 | 76 | |
| AK049680.1 | - | 8 - 9 | 1331 | 76 | ||
| AK088112.1 | - | 8 - 9 | 1328 | 76 | ||
| AK088671.1 | - | 9 - 10 | 1487 | 76 | ||
| AK133914.1 | - | 8 - 9 | 1326 | 76 | ||
| BC019412.1 | - | 8 - 9 | 1318 | 5151406 | 76 | |
| BC021152.1 | - | 8 - 9 | 1283 | 76 | ||
| BC043722.1 | - | 6 - 7 | 836 | 5321733 | 76 | |
| BC046814.1 | - | 8 - 9 | 1385 | 76 | ||
| CF738062.1 | - | 4 - 5 | 321 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Azin1
Genomic Map