Assay Details
| Gene Symbol: Tcerg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039474.1 | NP_001034563.1 | 10 - 11 | 1809 | 76 | |
| NM_001289526.1 | NP_001276455.1 | 9 - 10 | 1746 | 76 | ||
| NR_110344.1 | 10 - 11 | 1809 | 76 | |||
| GenBank mRNA | AB023485.1 | - | 10 - 11 | 1773 | 76 | |
| AK010571.1 | - | 9 - 10 | 1728 | 76 | ||
| AK076993.1 | - | 10 - 11 | 1788 | 76 | ||
| AK150312.1 | - | 9 - 10 | 1736 | 76 | ||
| AK168434.1 | - | 10 - 11 | 1787 | 76 | ||
| BC039185.1 | - | 9 - 10 | 1699 | 3982736 | 76 | |
| BC040284.1 | - | 9 - 10 | 1709 | 4460736 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcerg1
Genomic Map