Assay Details
| Gene Symbol: Nupr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019738.1 | NP_062712.1 | 1 - 2 | 150 | 117 | |
| GenBank mRNA | AF131196.1 | - | 1 - 2 | 150 | 117 | |
| AK009572.1 | - | 1 - 2 | 117 | |||
| BC002109.1 | - | 1 - 2 | 156 | 3487434 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupr1
Genomic Map