Assay Details
| Gene Symbol: Ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019826.3 | NP_062800.1 | 5 - 6 | 569 | 72 | |
| GenBank mRNA | AF225989.1 | - | 5 - 6 | 548 | 72 | |
| AK005024.1 | - | 5 - 6 | 588 | 72 | ||
| AK017660.1 | - | 5 - 6 | 555 | 72 | ||
| AK077416.1 | - | 5 - 6 | 576 | 72 | ||
| BC018325.1 | - | 5 - 6 | 559 | 4192286 | 72 | |
| BC027198.1 | - | 5 - 6 | 569 | 4237982 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ivd
Genomic Map