Assay Details
| Gene Symbol: Insm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020287.2 | NP_064683.2 | 1 | 1805 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452663 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452655 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452654 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452664 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452651 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452649 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452652 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452656 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058330 | NM_020287.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058335 | NM_020287.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119271 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058335 | NM_020287.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058330 | NM_020287.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P454542 | NM_020287.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28452669 | - | - |
| Whole Mouse Genome G4122A | A_51_P454542 | NM_020287.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09990636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09990641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09990646 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09990638 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452654 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452656 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452665 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452663 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452664 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452649 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452652 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452655 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28452651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28452667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09990642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09990641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09990638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09998054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09883657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09998057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09998047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10402990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10049302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09998049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10161788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452667 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395828 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437781_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453436_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422269_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395827 | - | - |
| Mouse Genome 430 2.0 Array | 1422269_at | - | - |
| Mouse Genome 430 2.0 Array | 1437781_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422269_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437781_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56856_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56856_PM_at | - | - |
| Mouse Expression Set 430 | 1422269_PM_at | - | - |
| Mouse Expression Set 430 | 1422269_at | - | - |
| Mouse Expression Set 430 | 1437781_PM_at | - | - |
| Mouse Expression Set 430 | 1437781_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422269_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422269_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422269_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422269_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453436_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437781_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453436_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437781_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 820022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724737 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724737 | - | - |
| mouse-6 v1.1 (Illumina) | scl3820.1.1_97-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl3820.1.1_97-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2891445 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2891445 | - | - |

Insm2
Genomic Map