Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193303.1 | NP_001180232.1 | 8 - 9 | 1020 | 63 | |
| NM_026148.3 | NP_080424.2 | 8 - 9 | 1014 | 63 | ||
| NM_201242.1 | NP_957694.1 | 8 - 9 | 1038 | 63 | ||
| GenBank mRNA | AK009390.1 | - | 8 - 9 | 1021 | 63 | |
| AK014954.1 | - | 8 - 9 | 1015 | 63 | ||
| AK082932.1 | - | 8 - 9 | 883 | 63 | ||
| AK088939.1 | - | 8 - 9 | 889 | 63 | ||
| AK151235.1 | - | 8 - 9 | 889 | 63 | ||
| AK151938.1 | - | 8 - 9 | 881 | 63 | ||
| AK152679.1 | - | 8 - 9 | 881 | 63 | ||
| AK153346.1 | - | 8 - 9 | 889 | 63 | ||
| BC005621.1 | - | 8 - 9 | 869 | 3711611 | 63 | |
| BC067393.1 | - | 8 - 9 | 812 | 63 | ||
| DQ003608.1 | - | 8 - 9 | 1005 | 63 | ||
| EU122953.1 | - | 8 - 9 | 1004 | 63 | ||
| EU122954.1 | - | 8 - 9 | 872 | 63 | ||
More Information
Set Membership: |
|

Lims1
Genomic Map