Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 5 - 6 | 506 | 81 | |
| NM_027328.4 | NP_081604.3 | 5 - 6 | 506 | 81 | ||
| GenBank mRNA | AK005294.1 | - | 5 - 6 | 474 | 81 | |
| AK033190.1 | - | 5 - 6 | 509 | 81 | ||
| AK033283.1 | - | 5 - 6 | 471 | 81 | ||
| AK044457.1 | - | 5 - 6 | 569 | 81 | ||
| AK051260.1 | - | 5 - 6 | 466 | 81 | ||
| AY040823.1 | - | 5 - 6 | 482 | 81 | ||
| BC018376.1 | - | 5 - 6 | 472 | 3673893 | 81 | |
| BC057877.1 | - | 4 - 5 | 449 | 5347609 | 81 | |
| BC061461.1 | - | 5 - 6 | 450 | 5717606 | 81 | |
| CT010189.1 | - | 4 - 5 | 426 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf31
Genomic Map