| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00017055.mm.1 |
NM_007515.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017055.mm.1 |
TC0X00002710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017058.mm.1 |
NM_007515.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017058.mm.1 |
TC0X00002710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017056.mm.1 |
NM_007515.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017056.mm.1 |
TC0X00002710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017060.mm.1 |
NM_007515.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017060.mm.1 |
TC0X00002710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017057.mm.1 |
NM_007515.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017057.mm.1 |
TC0X00002710.mm.1 |
MTA TC |