Assay Details
| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077593.1 | - | 3 - 4 | 668 | 75 | |
| AK141558.1 | - | 3 - 4 | 699 | 75 | ||
| AK165931.1 | - | 3 - 4 | 665 | 75 | ||
| BC050195.1 | - | 3 - 4 | 644 | 30010078 | 75 | |
| U70859.1 | - | 4 - 5 | 802 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a3
Genomic Map