Assay Details
| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077593.1 | - | 11 - 12 | 1862 | 111 | |
| AK141558.1 | - | 10 - 11 | 2334 | 111 | ||
| AK165931.1 | - | 11 - 12 | 1859 | 111 | ||
| BC050195.1 | - | 11 - 12 | 1838 | 30010078 | 111 | |
| U70859.1 | - | 12 - 13 | 1996 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988384 | BC050195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988384 | U70859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988384 | AK141558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988384 | NM_007515.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988384 | AK077593.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740156 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108254 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988384 | BC050195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988384 | U70859.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988384 | AK141558.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988384 | NM_007515.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988384 | AK077593.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275016 | BC050195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275016 | U70859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275016 | AK141558.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275016 | NM_007515.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275016 | AK077593.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32562407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108254 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09374950 | - | - |
| Whole Mouse Genome G4122A | A_51_P275016 | BC050195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275016 | U70859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275016 | AK141558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275016 | NM_007515.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275016 | AK077593.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122045 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132064 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14818131 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14516419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14516417 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14516421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32562324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14789800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14789808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14994957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14994946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14739325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15084600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14937340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15090933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14994954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15090950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14994948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15069116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14516421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32562370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14994963 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605997 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417022_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605986 | - | - |
| Mouse Genome 430 2.0 Array | 1417022_at | - | - |
| Mouse Genome 430 2.0 Array | 1417022_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11989_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11989_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00017037.mm.1 | NM_007515.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017037.mm.1 | TC0X00002710.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417022_at | - | - |
| Mouse Expression Set 430 | 1417022_PM_at | - | - |
| Murine Genome U74v2 Set | 93133_at | - | - |
| Murine Genome U74v2 Set | 93133_PM_at | - | - |
| Murine Genome U74v2 Set | 171155_i_at | - | - |
| Murine Genome U74v2 Set | 167669_i_at | - | - |
| Murine Genome U74v2 Set | 167669_PM_i_at | - | - |
| Murine Genome U74v2 Set | 171155_PM_i_at | - | - |
| Murine Genome U74v2 Set | 169264_r_at | - | - |
| Murine Genome U74v2 Set | 169264_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417022_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417022_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417022_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417022_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 60270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212906 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257987 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257987 | - | - |
| mouse-6 v1.1 (Illumina) | scl53996.13.1_171-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002937.1_42-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400091 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53996.13.1_171-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002937.1_42-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1212906 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257987 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257987 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1212906 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53996.13.1_171-S | - | - |

Slc7a3
Genomic Map