Assay Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008679.3 | NP_032705.2 | 13 - 14 | 2696 | 144 | |
| GenBank mRNA | AF000581.1 | - | 12 - 13 | 2588 | 144 | |
| AK137517.1 | - | 2 - 3 | 364 | 145 | ||
| AK142306.1 | - | 13 - 14 | 2601 | 144 | ||
| AK156345.1 | - | 13 - 14 | 2673 | 144 | ||
| BC086482.1 | - | 13 - 14 | 2666 | 30104508 | 144 | |
| BC093478.1 | - | 10 - 11 | 2327 | 144 | ||
| BC141177.1 | - | 12 - 13 | 2527 | 9055789 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa3
Genomic Map