Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018885.2 | NP_061373.1 | 1 - 2 | 383 | 54 | |
| GenBank mRNA | AF124732.1 | - | 1 - 2 | 88 | 54 | |
| AK132571.1 | - | 1 - 2 | 383 | 54 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF124732.1,AK132571.1,NM_018885.2 transcripts.
Set Membership: |
|

Irx4
Genomic Map