Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018885.2 | NP_061373.1 | 2 - 3 | 512 | 60 | |
| GenBank mRNA | AF124732.1 | - | 2 - 3 | 217 | 60 | |
| AK132571.1 | - | 2 - 3 | 512 | 60 | ||
| BC126942.1 | - | 2 - 3 | 150 | 40089986 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Irx4
Genomic Map