| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200039545.mm.1 |
NM_019696.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039545.mm.1 |
NM_019696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039545.mm.1 |
TC0200004623.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039544.mm.1 |
NM_019696.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039544.mm.1 |
NM_019696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039544.mm.1 |
TC0200004623.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039548.mm.1 |
NM_019696.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039548.mm.1 |
NM_019696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039548.mm.1 |
TC0200004623.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200039546.mm.1 |
NM_019696.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039546.mm.1 |
NM_019696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200039546.mm.1 |
TC0200004623.mm.1 |
MTA TC |