Assay Details
| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019696.2 | NP_062670.2 | 6 - 7 | 920 | 60 | |
| GenBank mRNA | AF077738.1 | - | 6 - 7 | 865 | 60 | |
| AK132611.1 | - | 2 - 3 | 166 | 60 | ||
| BC003713.1 | - | 6 - 7 | 853 | 3593211 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm1
Genomic Map