Assay Details
| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019696.2 | NP_062670.2 | 7 - 8 | 1012 | 89 | |
| GenBank mRNA | AF077738.1 | - | 7 - 8 | 957 | 89 | |
| AK132611.1 | - | 3 - 4 | 258 | 89 | ||
| BC003713.1 | - | 7 - 8 | 945 | 3593211 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm1
Genomic Map