Assay Details
| Gene Symbol: Cpxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019696.2 | NP_062670.2 | 9 - 10 | 1334 | 114 | |
| GenBank mRNA | AF077738.1 | - | 9 - 10 | 1279 | 114 | |
| AK132611.1 | - | 5 - 6 | 580 | 114 | ||
| BC003713.1 | - | 9 - 10 | 1267 | 3593211 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm1
Genomic Map