Assay Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021713.2 | NP_068359.1 | 5 - 6 | 791 | 80 | |
| GenBank mRNA | AF289484.1 | - | 5 - 6 | 783 | 80 | |
| AK002656.1 | - | 5 - 6 | 766 | 80 | ||
| AK004104.1 | - | 5 - 6 | 766 | 80 | ||
| AK013232.1 | - | 5 - 6 | 737 | 80 | ||
| BC023124.1 | - | 4 - 5 | 918 | 80 | ||
| BC028501.1 | - | 5 - 6 | 776 | 5256915 | 80 | |
More Information
Set Membership: |
|

Myg1
Genomic Map