Assay Details
| Gene Symbol: 0610012G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027897.1 | 1 | 880 | 72 | ||
| GenBank mRNA | AK002600.1 | - | 1 | 850 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417279 | NR_027897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417279 | AK002600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417279 | NR_027897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417279 | AK002600.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270869 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170312 | - | - |
| Whole Mouse Genome G4122A | A_51_P417279 | NR_027897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417279 | AK002600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417279 | NR_027897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417279 | AK002600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83419 | BC021536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83419 | AK009191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83419 | AK011122.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83419 | AK146135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83419 | NR_027897.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439019 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417002_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417003_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448540_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439018 | - | - |
| Mouse Genome 430 2.0 Array | 1448540_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417002_at | - | - |
| Mouse Genome 430 2.0 Array | 1417003_at | - | - |
| Mouse Genome 430 2.0 Array | 1417003_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417002_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448540_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106264_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106264_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600012249.mm.1 | NR_027897.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012249.mm.1 | NR_027897.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012249.mm.1 | TC1600001492.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600012250.mm.1 | NR_027897.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012250.mm.1 | NR_027897.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012250.mm.1 | TC1600001492.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600012251.mm.1 | NR_027897.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012251.mm.1 | NR_027897.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012251.mm.1 | TC1600001492.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448540_a_at | - | - |
| Mouse Expression Set 430 | 1448540_PM_a_at | - | - |
| Mouse Expression Set 430 | 1417002_at | - | - |
| Mouse Expression Set 430 | 1417002_PM_at | - | - |
| Mouse Expression Set 430 | 1417003_at | - | - |
| Mouse Expression Set 430 | 1417003_PM_at | - | - |
| Murine Genome U74v2 Set | 99166_at | - | - |
| Murine Genome U74v2 Set | 99166_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417002_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448540_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448540_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417003_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417003_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417002_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417002_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448540_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417003_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417003_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448540_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417002_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 433736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2658619 | - | - |

0610012G03Rik
Genomic Map