Assay Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159329.1 | NP_001152801.1 | 4 - 5 | 490 | 76 | |
| NM_025332.3 | NP_079608.1 | 4 - 5 | 691 | 76 | ||
| GenBank mRNA | AK002771.1 | - | 4 - 5 | 676 | 76 | |
| AK010979.1 | - | 4 - 5 | 688 | 76 | ||
| AK147840.1 | - | 4 - 5 | 675 | 76 | ||
| BC107216.1 | - | 4 - 5 | 688 | 40051675 | 76 | |
| BC107217.1 | - | 4 - 5 | 487 | 40051680 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtpbp8
Genomic Map