Assay Details
| Gene Symbol: Rrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025897.2 | NP_080173.1 | 4 - 5 | 1236 | 66 | |
| NM_133951.1 | NP_598712.1 | 4 - 5 | 1270 | 66 | ||
| GenBank mRNA | AK005138.1 | - | 4 - 5 | 1152 | 66 | |
| AK013462.1 | - | 4 - 5 | 1147 | 66 | ||
| AK045361.1 | - | 4 - 5 | 1088 | 66 | ||
| AK077786.1 | - | 4 - 5 | 1159 | 66 | ||
| AK154337.1 | - | 4 - 5 | 1263 | 66 | ||
| AK154858.1 | - | 4 - 5 | 1211 | 66 | ||
| AK165828.1 | - | 4 - 5 | 1084 | 66 | ||
| AK170257.1 | - | 4 - 5 | 1088 | 66 | ||
| AK172189.1 | - | 4 - 5 | 1109 | 66 | ||
| AK172224.1 | - | 4 - 5 | 1109 | 66 | ||
| BC022923.1 | - | 4 - 5 | 1168 | 4242458 | 66 | |
| BC046799.1 | - | 4 - 5 | 1143 | 6403599 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrp8
Genomic Map