Assay Details
| Gene Symbol: Rars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025936.3 | NP_080212.2 | 7 - 8 | 862 | 107 | |
| GenBank mRNA | AK011383.1 | - | 7 - 8 | 840 | 107 | |
| AK076160.1 | - | 7 - 8 | 837 | 107 | ||
| AK149856.1 | - | 7 - 8 | 837 | 107 | ||
| AK151966.1 | - | 6 - 7 | 660 | 107 | ||
| AK152108.1 | - | 7 - 8 | 837 | 107 | ||
| AK160216.1 | - | 7 - 8 | 837 | 107 | ||
| BC020132.1 | - | 7 - 8 | 820 | 3989952 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rars
Genomic Map