Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026824.4 | NP_081100.2 | 3 - 4 | 609 | 61 | |
| GenBank mRNA | AK004041.1 | - | 2 - 3 | 132 | 61 | |
| AK037538.1 | - | 2 - 3 | 1023 | 61 | ||
| AK088243.1 | - | 3 - 4 | 610 | 61 | ||
| AK147967.1 | - | 3 - 4 | 808 | 61 | ||
| AK148307.1 | - | 3 - 4 | 808 | 61 | ||
| AK183151.1 | - | 2 - 3 | 166 | 61 | ||
| AK184997.1 | - | 1 - 2 | 75 | 61 | ||
| AK185363.1 | - | 1 - 2 | 75 | 61 | ||
| AK211792.1 | - | 2 - 3 | 313 | 61 | ||
| BC019480.1 | - | 1 - 2 | 93 | 4037583 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004041.1,BC019480.1 transcripts.
Set Membership: |
|

Dus1l
Genomic Map