Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026824.4 | NP_081100.2 | 8 - 9 | 1104 | 64 | |
| GenBank mRNA | AK004041.1 | - | 7 - 8 | 627 | 64 | |
| AK037538.1 | - | 6 - 7 | 2241 | 64 | ||
| AK088243.1 | - | 8 - 9 | 1105 | 64 | ||
| AK147967.1 | - | 8 - 9 | 1304 | 64 | ||
| AK148307.1 | - | 8 - 9 | 1304 | 64 | ||
| BC019480.1 | - | 6 - 7 | 588 | 4037583 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dus1l
Genomic Map