Assay Details
| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027878.2 | NP_082154.2 | 5 - 6 | 723 | 102 | |
| GenBank mRNA | AK004552.1 | - | 5 - 6 | 723 | 102 | |
| AK151184.1 | - | 5 - 6 | 656 | 102 | ||
| BC021433.1 | - | 5 - 6 | 689 | 5030012 | 102 | |
More Information
Set Membership: |
|

Dram1
Genomic Map