| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800020919.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020919.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020919.mm.1 |
TC0800002729.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020921.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020921.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020921.mm.1 |
TC0800002729.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020920.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020920.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020920.mm.1 |
TC0800002729.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020924.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020924.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020924.mm.1 |
TC0800002729.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020917.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020917.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020917.mm.1 |
TC0800002729.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020922.mm.1 |
NM_028221.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020922.mm.1 |
NM_028221.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020922.mm.1 |
TC0800002729.mm.1 |
MTA TC |