Assay Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB120716.1 | - | 3 - 4 | 279 | 90 | |
| AK005610.1 | - | 3 - 4 | 279 | 90 | ||
| BC049742.1 | - | 2 - 3 | 223 | 6771761 | 90 | |
| JX089334.1 | - | 3 - 4 | 579 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata19
Genomic Map