Assay Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB120716.1 | - | 6 - 7 | 514 | 77 | |
| AK005610.1 | - | 6 - 7 | 514 | 77 | ||
| BC049742.1 | - | 5 - 6 | 458 | 6771761 | 77 | |
| JX089334.1 | - | 6 - 7 | 814 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067246 | BC049742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017535 | AB120716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017535 | AK005610.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017535 | NM_029299.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61665 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111427 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017535 | AB120716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017535 | AK005610.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017535 | NM_029299.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26375867 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01890947 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111427 | - | - |
| Whole Mouse Genome G4122A | A_51_P301007 | AB120716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301007 | AK005610.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301007 | BC049742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301007 | NM_029299.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106595 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479542 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479543 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07479541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26375847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07833599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08138844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07698439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07739910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08191666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07743119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07758155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08191664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07743111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07646930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07646925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07891779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07833598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08191673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26375834 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584017 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417863_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584015 | - | - |
| Mouse Genome 430 2.0 Array | 1417863_at | - | - |
| Mouse Genome 430 2.0 Array | 1417863_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75469_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75469_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900001987.mm.1 | NM_029299.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001987.mm.1 | TC0900000261.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417863_at | - | - |
| Mouse Expression Set 430 | 1417863_PM_at | - | - |
| Murine Genome U74v2 Set | 165474_f_at | - | - |
| Murine Genome U74v2 Set | 165474_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417863_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417863_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417863_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417863_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3140162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3140162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003140162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2636548 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2636548 | - | - |
| mouse-6 v1.1 (Illumina) | scl37179.8.1_83-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3140162 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37179.8.1_83-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2636548 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2636548 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37179.8.1_83-S | - | - |

Spata19
Genomic Map