Assay Details
| Gene Symbol: Pnpla3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054088.3 | NP_473429.2 | 4 - 5 | 756 | 61 | |
| GenBank mRNA | AY037763.1 | - | 4 - 5 | 736 | 61 | |
| BC028792.1 | - | 4 - 5 | 744 | 1265861 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla3
Genomic Map