Assay Details
| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080858.3 | NP_543134.1 | 1 - 2 | 140 | 78 | |
| GenBank mRNA | AF403040.1 | - | 1 - 2 | 141 | 78 | |
| AK009644.1 | - | 1 - 2 | 141 | 78 | ||
| BC107257.2 | - | 1 - 2 | 105 | 40052462 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF403040.1,AK009644.1,BC107257.2,NM_080858.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100360 | NM_080858.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51897 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143621 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100360 | NM_080858.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32541889 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03720035 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143621 | - | - |
| Whole Mouse Genome G4122A | A_51_P154524 | NM_080858.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154524 | AF403040.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154524 | AK009644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154524 | BC107257.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124708 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143621 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14834955 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14834946 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14834956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14834950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14834957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32541915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14676097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14569887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14836271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15135664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15038852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15225901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14763846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14667497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14987763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15038853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15171472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15038846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32541897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611731 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605817 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420346_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605815 | - | - |
| Mouse Genome 430 2.0 Array | 1420346_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420346_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70392_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70392_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00016689.mm.1 | NM_080858.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016689.mm.1 | NM_080858.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016689.mm.1 | TC0X00002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00016687.mm.1 | NM_080858.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016687.mm.1 | NM_080858.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016687.mm.1 | TC0X00002668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00016688.mm.1 | NM_080858.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016688.mm.1 | NM_080858.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016688.mm.1 | TC0X00002668.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420346_PM_at | - | - |
| Mouse Expression Set 430 | 1420346_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420346_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420346_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420346_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420346_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 494017 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7040398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714497 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2714497 | - | - |
| mouse-6 v1.1 (Illumina) | scl54027.6.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54027.6.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2965123 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2965123 | - | - |

Asb12
Genomic Map