Assay Details
| Gene Symbol: Trhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133202.2 | NP_573465.1 | 2 - 3 | 805 | 72 | |
| GenBank mRNA | AF283762.1 | - | 2 - 3 | 805 | 72 | |
| BC117988.2 | - | 2 - 3 | 784 | 40104412 | 72 | |
| BC117989.2 | - | 2 - 3 | 784 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985773 | NM_133202.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102895 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161209 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985773 | NM_133202.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P352940 | BC117988.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P352940 | BC117989.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P352940 | AF283762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P352940 | NM_133202.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26232890 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161209 | - | - |
| Whole Mouse Genome G4122A | A_51_P352940 | BC117988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P352940 | BC117989.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P352940 | AF283762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P352940 | NM_133202.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122487 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06686222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07233553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07233548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07233550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06686228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26232865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07327375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07327364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06759107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06686220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07327368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07415217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06686222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07327372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07415223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06796194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07327370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07330602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07050476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07330605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07233548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06686227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26232884 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10582301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582302 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450356_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10582299 | - | - |
| Mouse Genome 430 2.0 Array | 1450356_at | - | - |
| Mouse Genome 430 2.0 Array | 1450356_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170732_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170732_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800023672.mm.1 | NM_133202.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023672.mm.1 | NM_133202.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023672.mm.1 | TC0800003066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800023671.mm.1 | NM_133202.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023671.mm.1 | NM_133202.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800023671.mm.1 | TC0800003066.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450356_PM_at | - | - |
| Mouse Expression Set 430 | 1450356_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450356_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450356_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450356_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450356_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 760166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 110398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2461612 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2461612 | - | - |
| mouse-6 v1.1 (Illumina) | scl34229.3.1_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34229.3.1_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2461612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2461612 | - | - |

Trhr2
Genomic Map