Assay Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133683.3 | NP_598444.1 | 5 - 6 | 872 | 67 | |
| GenBank mRNA | AK018383.1 | - | 5 - 6 | 820 | 67 | |
| AK034298.1 | - | 5 - 6 | 878 | 67 | ||
| AK036228.1 | - | 4 - 5 | 949 | 67 | ||
| AK042426.1 | - | 4 - 5 | 927 | 67 | ||
| AK079735.1 | - | 730 | 67 | |||
| AK136778.1 | - | 862 | 67 | |||
| AK138931.1 | - | 5 - 6 | 860 | 67 | ||
| AK155275.1 | - | 5 - 6 | 833 | 67 | ||
| AK170143.1 | - | 5 - 6 | 832 | 67 | ||
| AK171087.1 | - | 5 - 6 | 832 | 67 | ||
| BC016895.1 | - | 5 - 6 | 874 | 4239007 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem19
Genomic Map