Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133697.3 | NP_598458.1 | 1 - 2 | 102 | 135 | |
| GenBank mRNA | AK018994.1 | - | 1 - 2 | 111 | 135 | |
| AK044669.1 | - | 1 - 2 | 206 | 135 | ||
| AK088833.1 | - | 1 - 2 | 149 | 135 | ||
| AK164394.1 | - | 1 - 2 | 103 | 135 | ||
| BC009097.1 | - | 1 - 2 | 97 | 3481841 | 135 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK018994.1,AK044669.1,AK088833.1,AK164394.1,BC009097.1,NM_133697.3 transcripts.
Set Membership: |
|

Smim14
Genomic Map