Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133697.3 | NP_598458.1 | 2 - 3 | 206 | 75 | |
| GenBank mRNA | AK003345.1 | - | 2 - 3 | 201 | 75 | |
| AK018994.1 | - | 2 - 3 | 215 | 75 | ||
| AK044669.1 | - | 2 - 3 | 310 | 75 | ||
| AK088833.1 | - | 2 - 3 | 253 | 75 | ||
| AK164394.1 | - | 2 - 3 | 207 | 75 | ||
| BC009097.1 | - | 2 - 3 | 201 | 3481841 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smim14
Genomic Map