Assay Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133816.2 | NP_598577.1 | 2 - 3 | 217 | 65 | |
| GenBank mRNA | AK028826.1 | - | 2 - 3 | 197 | 65 | |
| AK032634.1 | - | 2 - 3 | 258 | 65 | ||
| AK041504.1 | - | 2 - 3 | 265 | 65 | ||
| BC012284.1 | - | 2 - 3 | 217 | 3978494 | 65 | |
| BC034733.1 | - | 1 - 2 | 53 | 4947387 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028826.1,AK032634.1,AK041504.1,BC012284.1,BC034733.1,NM_133816.2 transcripts.
Set Membership: |
|

Sh3bp4
Genomic Map