Assay Details
Gene Symbol: Atp6v1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133826.4 | NP_598587.2 | 1 - 2 | 101 | 90 | |
GenBank mRNA | AK034165.1 | - | 1 - 2 | 151 | 90 | |
AK078767.1 | - | 1 - 2 | 102 | 90 | ||
AK081492.1 | - | 1 - 2 | 111 | 90 | ||
AK135137.1 | - | 1 - 2 | 143 | 90 | ||
AK151659.1 | - | 1 - 2 | 164 | 90 | ||
AK152147.1 | - | 1 - 2 | 122 | 90 | ||
BC009154.1 | - | 1 - 2 | 102 | 3601621 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK078767.1,AK151659.1,BC009154.1,NM_133826.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
