Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293694.1 | NP_001280623.1 | 5 - 6 | 909 | 132 | |
| GenBank mRNA | BC021753.1 | - | 5 - 6 | 838 | 4167222 | 132 |
More Information
Updated Mapping Information
Set Membership: |
|

Cept1
Genomic Map