Assay Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134093.2 | NP_598854.1 | 1 - 2 | 211 | 131 | |
| GenBank mRNA | AF287293.1 | - | 1 - 2 | 151 | 131 | |
| AK046496.1 | - | 1 - 2 | 131 | 131 | ||
| AK080221.1 | - | 1 - 2 | 131 | 131 | ||
| AK154676.1 | - | 1 - 2 | 126 | 131 | ||
| BY293457.1 | - | 1 - 2 | 211 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Letmd1
Genomic Map