Assay Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134093.2 | NP_598854.1 | 4 - 5 | 565 | 117 | |
| GenBank mRNA | AF287293.1 | - | 4 - 5 | 505 | 117 | |
| AF401483.1 | - | 4 - 5 | 398 | 117 | ||
| AK041312.1 | - | 3 - 4 | 894 | 117 | ||
| AK045256.1 | - | 4 - 5 | 385 | 117 | ||
| AK046496.1 | - | 3 - 4 | 369 | 117 | ||
| AK080221.1 | - | 4 - 5 | 485 | 117 | ||
| AK154676.1 | - | 4 - 5 | 480 | 117 | ||
| AK158718.1 | - | 3 - 4 | 342 | 117 | ||
| AK169341.1 | - | 4 - 5 | 485 | 117 | ||
| BC021361.1 | - | 1 - 2 | 237 | 5010892 | 117 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC021361.1 transcripts.
Set Membership: |
|

Letmd1
Genomic Map