Assay Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134096.2 | NP_598857.1 | 1 - 2 | 256 | 62 | |
| GenBank mRNA | AY325124.1 | - | 1 - 2 | 257 | 62 | |
| BC015306.1 | - | 1 - 2 | 257 | 4223181 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam19a5
Genomic Map