| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_001161355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_001161355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_001161356.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_001161356.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_134249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
NM_134249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024038.mm.1 |
TC1100002611.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_001161355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_001161355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_001161356.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_001161356.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_134249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
NM_134249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024039.mm.1 |
TC1100002611.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_001161355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_001161355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_001161356.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_001161356.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_134249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
NM_134249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024036.mm.1 |
TC1100002611.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_001161355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_001161355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_001161356.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_001161356.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_134249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
NM_134249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024037.mm.1 |
TC1100002611.mm.1 |
MTA TC |