Assay Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144784.3 | NP_659033.1 | 6 - 7 | 643 | 89 | |
| GenBank mRNA | AJ510150.1 | - | 6 - 7 | 635 | 89 | |
| AK032097.1 | - | 6 - 7 | 614 | 89 | ||
| AK075593.1 | - | 6 - 7 | 616 | 89 | ||
| AK081715.1 | - | 6 - 7 | 643 | 89 | ||
| AK146241.1 | - | 6 - 7 | 598 | 89 | ||
| AK163402.1 | - | 6 - 7 | 647 | 89 | ||
| AK169771.1 | - | 6 - 7 | 636 | 89 | ||
| BC024429.1 | - | 6 - 7 | 646 | 89 | ||
| BC024763.1 | - | 6 - 7 | 615 | 5365469 | 89 | |
| BC029345.1 | - | 3 - 4 | 304 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acat1
Genomic Map