| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900016538.mm.1 |
NM_001102455.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016538.mm.1 |
NM_001102455.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016538.mm.1 |
TC0900001979.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_001102455.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_001102455.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_001102456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_001102456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_009691.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
NM_009691.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016535.mm.1 |
TC0900001979.mm.1 |
MTA TC |