Assay Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102455.1 | NP_001095925.1 | 9 - 10 | 1385 | 83 | |
NM_001102456.1 | NP_001095926.1 | 8 - 9 | 1217 | 83 | ||
NM_009691.2 | NP_033821.1 | 8 - 9 | 1217 | 83 | ||
GenBank mRNA | AK149977.1 | - | 9 - 10 | 1386 | 83 | |
AK150020.1 | - | 9 - 10 | 1386 | 83 | ||
AK150478.1 | - | 1 - 2 | 117 | 83 | ||
AK151708.1 | - | 1 - 2 | 117 | 83 | ||
AK152787.1 | - | 1 - 2 | 117 | 83 | ||
AK153087.1 | - | 1 - 2 | 117 | 83 | ||
AK164012.1 | - | 9 - 10 | 1403 | 83 | ||
AK200131.1 | - | 2 - 3 | 133 | 83 | ||
BC051999.1 | - | 8 - 9 | 1195 | 5716148 | 83 | |
BC052396.1 | - | 8 - 9 | 1189 | 5719918 | 83 | |
M97216.1 | - | 4 - 5 | 577 | 83 | ||
U15571.1 | - | 9 - 10 | 1361 | 83 | ||
Z22592.1 | - | 8 - 9 | 1217 | 83 | ||
Z27070.1 | - | 8 - 9 | 1215 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK150478.1, AK151708.1, AK152787.1, AK153087.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
